Learn More
Cayman Chemical FTY720 phenoxy-bIotIn 100ug
SDP
Supplier:  Cayman Chemical 13254100 UG
Encompass_Preferred
A biotin-tagged analog of FTY720; methanol side chain of FTY720 that is targeted for phosphorylation by sphingosine kinases is retained, suggesting that it would likewise be phosphorylated in vivo
Catalog No. 50-295-2868