Learn More
Cayman Chemical FTY720 Phosphate, 402615-91-2, 5 mg
SDP
Supplier:  Cayman Chemical 100086395
Encompass_Preferred
Molecular Formula C19H34NO5P, Purity ≥98%, Formula Weight 387.5, Synonyms: FTY720P; A potent agonist at four of the sphingosine-1-phosphate (S1P) receptors with IC50 values of 0.2-6 nM.
Catalog No. NC1513125