Learn More
Medchemexpress LLC FUBP1-IN-1 | 883003-62-1 | 99.7% | 405.39 | C19H14F3N3O2S | 100 MG

Supplier: Medchemexpress LLC HY100758100MG
A small-molecule inhibitor of FUSE binding protein 1 (FUBP1) that disrupts FUBP1 binding to single-stranded FUSE DNA; reported IC50 11.0 μM. Supplied as a light yellow solid for research use, characterized by high purity and defined storage and solvent recommendations.
- Potent FUBP1 inhibitor with an IC50 of 11.0 μM.
- High purity (99.72%).
- Molecular formula C19H14F3N3O2S and molecular weight 405.39.
- Supplied as a 100 MG solid with good solubility in DMSO (16.67 mg/mL).
- Recommended solvent DMSO for stock solutions.
- Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (2 years) or -20°C (1 year).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.