Learn More
Medchemexpress LLC Fubp1-in-1 | 883003-62-1 | 99.7% | 405.39 g/mol | C19H14F3N3O2S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10075825MG
Encompass_Preferred
FUBP1-IN-1 is a small-molecule inhibitor of FUSE binding protein 1 (FUBP1) that disrupts FUBP1 binding to single-stranded FUSE DNA. It is intended for use in biochemical and cellular research to probe FUBP1-related pathways and for target validation and mechanistic studies.
  • Potent FUBP1 inhibitor (reported IC50 11.0 μM).
  • High purity suitable for research applications (99.7%).
  • Well characterized: molecular weight 405.39 g/mol; formula C19H14F3N3O2S.
  • Available in small-scale quantities suitable for laboratory testing.
  • Useful for target validation and mechanistic studies in cellular and biochemical assays.
Catalog No. 50-002-13611