Learn More
Medchemexpress LLC FUBP1-IN-1 | 883003-62-1 | 99.7% | 405.39 g·mol⁻¹ | C19H14F3N3O2S | 50 MG

Supplier: Medchemexpress LLC HY10075850MG
FUBP1-IN-1 is a small-molecule inhibitor of far upstream element binding protein 1 (FUBP1) that interferes with FUBP1 binding to single-stranded FUSE DNA. It is characterized by an IC50 of 11.0 μM and is supplied as a high-purity research reagent for biochemical and cellular studies.
- Inhibits FUBP1 with an IC50 of 11.0 μM.
- High purity (≈99.7%).
- Molecular formula C19H14F3N3O2S; molecular weight 405.39 g·mol⁻¹.
- Available as solid vials (including 50 mg) and as a 10 mM solution in DMSO.
- Powder stability at -20°C for long term; in-solution storage at -80°C recommended.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.