Learn More
GENOVIS INC FucosEXO, 2000 units, Origin: E. Coli
Supplier:  GENOVIS INC G1FM1020
Encompass
FucosEXO is a mix of a-fucosidases for efficient hydrolysis of a1-2, a1-3 and a1-4-linked fucose on N- and O-glycosylated proteins or free oligosaccharides. FucosEXO is available as a 2000 units lyophilized fucosidase mix for hydrolysis of fucose from 2 mg glycoprotein.
Catalog No. NC1943556