Learn More
Medchemexpress LLC Fucoxanthin | 3351-86-8 | 1 ML

Supplier: Medchemexpress LLC HYN230210MM/1ML
Fucoxanthin is a marine carotenoid that exhibits various biological activities, including anti-obesity, anti-diabetic, anti-oxidant, anti-inflammatory, and anticancer properties. This product is supplied as a 10 mM solution in 1 mL of DMSO.
- High purity (99.38%)
- Molecular weight: 658.91
- Chemical formula: C42H58O6
- Targets multiple pathways, including PPARγ, UCP1, and caspases
- Shipped at room temperature
- Store at -80°C for up to 6 months or -20°C for up to 1 month when in solvent
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.