Learn More
Apexbio Technology LLC Fudosteine 13189-98-5 10mM (in 1mL H2O)
SDP
Supplier:  Apexbio Technology LLC B17635.1
Encompass_Preferred
Fudosteine (13189-98-5) is a cysteine-derived small molecule targeting MUC5AC mucin gene expression It is designed to modulate mucus secretion by airway epithelial cells thereby regulating excessive mucin production in the respiratory tract Fudosteine exerts its biological activity primarily by inhibiting MUC5AC gene expression In cell-based and animal disease models Fudosteine demonstrates inhibitory activity on mucin synthesis Based on these pharmacological properties Fudosteine holds research potential in investigating mechanisms underlying mucin synthesis control mucus-associated respiratory pathology and potential therapeutic targets for respiratory disorders
Catalog No. 50-000-94140