Learn More
Cole-Parmer PVC Fuel and Lubricant Tubing, Yellow, 7/16" ID x 9/16" OD; 25 Ft
Supplier:  Cole-Parmer 5012140
Encompass_Preferred
Cole-Parmer PVC Fuel and Lubricant tubing is the ideal transfer tubing for kerosene, gasoline, oils, cutting compounds, and other chemicals and petrochemicals. This tubing is formulated to resist the corrosive effects of hydrocarbon-based fluids, avoid swelling and hardening, and protect against product contamination and leaching. These fuel line hoses are flexible enough to allow for fast, simple installation in even the tightest areas, and maintain this flexibility over time to reduce failure due to cracking and leaking. This fuel tubing’s transparent yellow color provides for easy identification and flow monitoring, and it is available in the most common sizes to suit nearly any need involving fuel and lubricant applications.
Catalog No. 50-255-7694