Learn More
Gilson Company Inc 8" All Stainless Sieve Pan, Full Height
Supplier:  Gilson Company Inc V8SFXPN
Encompass_Preferred
Sieve Pan, 8in (203mm) diameter Stainless Steel full height Pan for ASTM E11 8in Test Sieves is placed at the bottom of a sieve stack to collect fines passing through the bottom sieve.
Catalog No. NC0576639