Learn More
Medchemexpress LLC Fullerene-C60 | 99685-96-8 | MFCD00151408 | 99.7% | 720.64 | C60 | 5 MG

Supplier: Medchemexpress LLC HYW0207685MG
Fullerene-C60 is the C60 carbon allotrope used in photodynamic research, nanomaterials studies, and biochemical assays. This high-purity solid powder is provided for research applications and exhibits limited solubility in aqueous media.
- High purity (99.73%).
- Chemical formula C60 and molecular weight 720.64 g/mol.
- Solid powder appearance suitable for analytical and materials research.
- Low solubility in water (<0.1 mg/mL) and limited solubility in DMSO (<1 mg/mL).
- Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
- Suitable for photodynamic and nanomaterials applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.