Learn More
Medchemexpress LLC Fulvestrant-d5 (ICI 182780-d5) | 00-00-0 | 99.7% | 611.80 | C32H42D5F5O3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY13636S15MG
Encompass_Preferred
Fulvestrant-d5 is a deuterium-labeled analogue of fulvestrant used as an internal standard in bioanalytical and mass spectrometry workflows. It retains the parent compound's structure while providing a distinct mass shift for selective detection and precise quantitation in pharmacokinetic, metabolic, and residue analysis.
  • Deuterium-labeled internal standard for LC-MS/MS assays.
  • Enables accurate quantitation and reduced matrix effects.
  • Suitable for pharmacokinetic and metabolic studies.
  • High chemical purity and defined storage stability.
Catalog No. 50-002-22247