Learn More
AdipoGen Fulvic Acid (5 mg)
SDP
Supplier:  AdipoGen AGCN20135M005
Encompass_Preferred
Chemical. CAS: 479-66-3. Formula: C14H12O8. MW: 308.2. Isolated from Penicillium sp. strain FKP-0046. Free radical scavenger and antioxidant. Potential compound to neutralize radioactive and toxic wastes and "heal" soils. Chelates and binds scores of minerals into a bioavailable form used by cells. Shown to incorporate into bone and cartilage of rats. Antibacterial. Antifungal. beta-Hexosaminidase release inhibitor. Anti-inflammatory. Tau fibrils aggregation inhibitor (Alzheimers Disease). Antiseptic. Interrupts the dimer formation of Abeta(17-42) peptides.
Catalog No. 50-205-3510