Learn More
Medchemexpress LLC Fumarate hydratase-I 25mg
Supplier: Medchemexpress LLC HY10000425MG
Fumarate hydratase-IN-1 (compound 2) is a cell-permeable fumarate hydratase inhibitor Fumarate hydratase-IN-1 has antiproliferative activity against several cancer cell lines with a mean IC50 of 2 2 M1]