Learn More
Medchemexpress LLC HY-100004 2mg Medchemexpress, Fumarate hydratase-IN-1 CAS:1644060-37-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1000042MG
Encompass_Preferred
Medchemexpress, HY-100004 2mg Fumarate hydratase-IN-1 CAS:1644060-37-6 Fumarate hydratase-IN-1 (compound 3) is a cell-permeable fumarate hydratase inhibitor. Fumarate hydratase-IN-1 has antiproliferative activity against several cancer cell lines with a mean IC 50 of 2.2 μM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1682