Learn More
Medchemexpress LLC HY-100005A 5mg Medchemexpress, Fumarate hydratase-IN-2 (sodium salt) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100005A5MG
Encompass_Preferred
Medchemexpress, HY-100005A 5mg Fumarate hydratase-IN-2 (sodium salt) CAS: Fumarate hydratase-IN-2 sodium salt (compound 3) is a cell-permeable and competitive fumarate hydratase inhibitor ( K i =4.5 μM) with nutrient-dependent cytotoxicity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1683