Learn More
Medchemexpress LLC Fumaric acid-13C2,d2 | 100858-52-4 | 98.0% | 120.07 g/mol | C213C2H2D2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW015883S25MG
Encompass_Preferred
Fumaric acid-13C2,d2 is a stable isotope-labeled fumaric acid supplied as a high-purity, white to off-white solid for tracer and analytical applications. Labeled with two 13C atoms and two deuteriums, it is intended for use in mass spectrometry standards, metabolic studies, and analytical method development where isotopic enrichment is required.
  • Stable isotope labeled with 13C2 and d2 for tracer studies.
  • High purity (98.0%) suitable for analytical applications.
  • White to off-white solid, easy to handle and weigh.
  • Useful as an internal standard for mass spectrometry and metabolic tracing.
  • Available in small research-scale quantities for method development.
Catalog No. 50-003-00556