Learn More
Medchemexpress LLC Fumaric acid-d2 | 24461-32-3 | MFCD00144378 | 99.0% | 118.08 g/mol | C4H2D2O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW015883S310MG
Encompass_Preferred
Fumaric acid-d2 is the deuterium-labeled isotopologue of fumaric acid used as a stable isotope reference standard and tracer for analytical and metabolic research. Supplied as a high-purity solid, it is suitable for mass spectrometry and NMR applications and is provided in small laboratory pack sizes for convenience.
  • High purity suitable for analytical use.
  • Deuterium-labeled (d2) isotopologue for tracer studies.
  • White to off-white solid powder with specified storage conditions.
  • Soluble in DMSO at ≥100 mg/mL for solution preparation.
  • Available in multiple small pack sizes for laboratory workflows.
Catalog No. 50-003-00557