Learn More
Medchemexpress LLC Fumaric acid-d4 | 194160-45-7 | MFCD00190450 | 98.0% | 120.10 | C4D4O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW015883S410MG
Encompass_Preferred
Fumaric acid-d4 is the deuterium-labeled form of fumaric acid used as a stable isotope tracer and internal standard for quantitative and metabolic studies. It is intended for research use and is commonly applied in analytical workflows where isotopic labeling improves detection, tracking, and quantitation of fumaric acid-related metabolites.
  • Deuterium labeled for tracer and internal standard applications.
  • Suitable for NMR, GC-MS, and LC-MS quantitation.
  • High purity (98%) for analytical workflows.
  • Solid powder with recommended low-temperature storage.
  • Available in small, research-scale sizes for analytical use.
Catalog No. 50-003-00983