Learn More
Medchemexpress LLC Fumaric acid-d4 | 194160-45-7 | MFCD00190450 | 98.0% | 120.10 | C4D4O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW015883S45MG
Encompass_Preferred
Deuterium-labeled fumaric acid (fumaric acid-d4) is an isotopically labeled reference material for use in metabolomics, tracer experiments, and analytical method development. It provides a characterized standard for quantitation and pathway studies involving fumaric acid and related metabolites.
  • HPLC purity 98%.
  • Molecular formula C4D4O4; molecular weight 120.10.
  • Suitable for metabolomics and tracer studies.
  • Supplied as a small-mass solid for analytical use.
  • Certificate of analysis available for batch-specific validation.
Catalog No. 50-003-00272