Learn More
HCL Labels HCL Labels Keep Fume Hood Sash Closed Energy Conservation Label - 1.25" x 20" Adhesive Vinyl Sign
SDP
Supplier:  HCL Labels SSLA114
Encompass_Preferred
An effective way to improve chemical fume hood's safety for less operating cost.This updated design uses the color scheme developed by the Danish Technical University in Copenhagen, with text to deliver a clear message: “Safer - Less Energy!”
Catalog No. NC1974409