Learn More
Cayman Chemical FumIquInazolIn D 5mg
SDP
Supplier:  Cayman Chemical 238485 MG
Encompass_Preferred
A fungal metabolite with antibacterial and antifungal properties; active against Gram-positive and Gram-negative bacteria (MICs = 8-16 UG/ml); active against F. solani and C. albicans fungi (MICs = 32 and 64 UG/ml)
Catalog No. 50-299-4801