Learn More
Cayman Chemical FumonIsIn B3 1mg
SDP
Supplier:  Cayman Chemical 204341 MG
Encompass_Preferred
A mycotoxin; cytotoxic to primary rat hepatocytes at 125, 250, and 500 µM; induces hepatocyte nodules in rats at 0.05% in the diet for 14 or 21 days; has been detected in corn and corn-based foods and livestock feeds
Catalog No. 50-298-2319