Learn More
Cayman Chemical FungI Asprgls KotnIn A 2.5mg
SDP
Supplier:  Cayman Chemical 249172.5 MG
Encompass_Preferred
A fungal metabolite; produced by A. glaucus (A. clavatus), A. alliaceus, and A. niger; toxic to day-old chicks (LD100 = 62.5 mg/kg)
Catalog No. 50-293-3791