Learn More
Cayman Chemical FungI FusarIum EnIatIn B 5mg
SDP
Supplier:  Cayman Chemical 153825 MG
Encompass_Preferred
Inhibits the Pdr5p in yeast, augmenting, at concentrations as low as 0.8 μM, the ability of cerulenin or cycloheximide to impair cell proliferation; inhibits ACAT (IC50 = 113 μM), blocking cholesteryl ester formation
Catalog No. 50-296-9097