Learn More
Fritsch Milling And Sizing Inc Funnel 5 mm for Wet Dividing, L-27
SDP
Supplier:  Fritsch Milling And Sizing Inc 27.1330.16
Encompass
Funnel 5 mm for division of suspensions for LABORETTE 27 with dividing head, division ratio 1:30
. Funnels are also available in 5, 15 and 22 mm dia. to suit the particle size of the sample being divided.
Catalog No. 50-153-4077