Learn More
Wilmad Labglass FUNNEL ADD CYL GRAD 125ML
Supplier:  Wilmad Labglass LG8271T102
Encompass
Funnel Addition, Graduated, Pressure Equalizing, PTFE Stopcock. Capacity: 125mL; Subdivisions: 1mL; Joint Size: 24/40; Stopcock Bore: 2. Addition funnel w/ double Scale graduations.
Catalog No. 50-806-664