Learn More
Wilmad Labglass FUNNEL ADD CYL GRAD 125ML
Supplier:  Wilmad Labglass LG8273T102
Encompass
Funnel Addition, Graduated, Pressure Equalizing, PTFE Stopcock. Capacity: 125mL; Subdivisions: 1mL; Joint Size: 24/40; Stopcock Size: 2. Addition funnel w/ double Scale graduations and stopcock in the pressure equalizing sidearm.
Catalog No. 50-805-206