Learn More
Wilmad Labglass FUNNEL ADD CYL GRAD 500ML
Supplier:  Wilmad Labglass LG8273T106
Encompass
Funnel Addition, Graduated, Pressure Equalizing, PTFE Stopcock. Capacity: 500mL; Subdivisions: 5mL; Joint Size: 24/40; Stopcock Size: 4. Addition funnel w/ double Scale graduations and stopcock in the pressure equalizing sidearm.
Catalog No. 50-804-154