Learn More
Wilmad Labglass FUNNEL BUCHNER V/ADP 600ML M
Supplier:  Wilmad Labglass LG7120129
Encompass_Preferred
Filter Funnel Buchner w/ Vacuum Adapter. Capacity: 600mL. Disc Porosity: Mediume. Joint Size: [ST] 24/40 Inner. Buchner filter funnel w/ a sealed-in fritted Glass disc w/ the listed porosity.
Catalog No. 50-803-905