Learn More
Wilmad Labglass FUNNEL PWDR 60O 100MM 24/40
Supplier:  Wilmad Labglass LG8305102
Encompass_Preferred
Funnel Powder, 60degree. Approximate Diameter at Top: 100mm; Joint Size: [ST] 24/40 Inner. For introduction of liquids or powders.
Catalog No. 50-807-675