Learn More
Wilmad Labglass FUNNEL PWDR OFFSET 75MM 24/40
Supplier:  Wilmad Labglass LG8303100
Encompass_Preferred
Funnel, Powder; Wilmad LabGlass; Offset with one straight side; Used with multi neck flasks; For introduction of liquids or powders into containers; Top diameter: 75mm; Joint Size: 24/50
Catalog No. 50-806-351
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.