Learn More
Wilmad Labglass FUNNEL SEP SQB 125ML 24/40
Supplier:  Wilmad Labglass LG8431T102
Encompass_Preferred
Separatory Funnel, Squibb, PTFE Stopcock. Capacity: 125mL. Lower Joint Size: [ST] 24/40. Stopcock Size: 2. Stopper Size: 22. Squibb separatory funnel with PTFE Stopcock and lower standard taper inner joint with drip tip.
Catalog No. 50-807-212