Learn More
Wilmad Labglass FUNNEL SEP SQB 500ML POLY NO27
Supplier:  Wilmad Labglass LG8422T106
Encompass_Preferred
Separatory Funnel, Squibb, PTFE Stopcock. Capacity: 500mL. Stopcock Size: 4. Stopper Size: 27. Squibb separatory funnel with polytetrafluoroethylene (PTFE) Stopcock. Supplied complete with a plastic flask length Stopper of the size listed.
Catalog No. 50-803-839