Learn More
Wilmad Labglass FUNNEL SEP SQB ECON 125ML
Supplier:  Wilmad Labglass LG8435T102
Encompass_Preferred
Separatory Funnel, Squibb, Detachable PTFE Stopcock. Capacity: 125mL. Stopper Size: 22. Economy style Squibb separatory funnel has a detachable polytetrafluoroethylene (PTFE) Stopcock and glass drip tip.
Catalog No. 50-806-521