Learn More
Wilmad Labglass FUNNEL SEP SQB ECON 500ML
Supplier:  Wilmad Labglass LG8435T106
Encompass_Preferred
Separatory Funnel, Squibb, Detachable PTFE Stopcock. Capacity: 500mL. Stopper Size: 27. Economy style Squibb separatory funnel has a detachable polytetrafluoroethylene (PTFE) Stopcock and glass drip tip.
Catalog No. 50-804-263