Learn More
Wilmad Labglass FUNNEL SEP SQB PTFE S/C 1000ML
Supplier:  Wilmad Labglass LG8421T110
Encompass_Preferred
Separatory Funnel, Squibb, PTFE Stopcock. Capacity: 1000mL. Stopcock Size: 4. Stopper Size: 27. Squibb separatory funnel with polytetrafluoroethylene (PTFE) Stopcock. Supplied complete with glass Pennyhead flask length Stopper.
Catalog No. 50-804-134