Learn More
Wilmad Labglass FUNNEL SEP SQB PTFE S/C 10ML
Supplier:  Wilmad Labglass ML1441T700
Encompass_Preferred
Separatory Funnel, Squibb, PTFE Stopcock. Capacity: 10mL. Stopcock Size: 2. Stopper Size: 14/20. Supplied complete with glass Pennyhead flask length Stopper of the size listed.
Catalog No. 50-807-154