Learn More
Wilmad Labglass FUNNEL SEP SQB PTFE S/C 3000ML
Supplier:  Wilmad Labglass LG8421T114
Encompass_Preferred
Separatory Funnel, Squibb, PTFE Stopcock. Capacity: 3000mL. Stopcock Size: 8. Stopper Size: 38. Squibb separatory funnel with polytetrafluoroethylene (PTFE) Stopcock. Supplied complete with glass Pennyhead flask length Stopper.
Catalog No. 50-806-554