Learn More
Chemglass Life Sciences Funnel, Vial, PTFE, 1.0" OD, 8-425 GPI Thread
SDP
Supplier:  Chemglass Life Sciences CG176101
Encompass_Preferred
Funnel, Vial, PTFE, 1.0" OD, 8-425 GPI Thread. Funnel machined from PTFE screws securely onto any standard laboratory vial having a GPI thread. Design allows for easy, safe and mess free transfer of product.
Catalog No. 50-153-1229