Learn More
Bellco Glass, Inc. Funnels. – 60mm, Polypropylene
SDP
Supplier:  Bellco Glass, Inc. 130000218
Encompass_Preferred
Funnels feature internal ribs to promote faster flow and external ribs to help prevent airlock.Polystyrene funnels are appropriate for use with mild acidic or alkali solutionsPolypropylene funnels are suitable for use with organic solvents and stronger acids and alkalisFunnels measure approximately 60 mm (2½") wide at the top with a 60° taper to the funnel stem. Material-Polypropylene (PP), Color-Natural, Diameter (mm)-60, Height (mm)-81.2, Package Type- bulk, Carton Qty-100
Catalog No. 50-233-0522