Learn More
Wilmad Labglass FUNNL SEP SQB PTFE 125ML 19/22
Supplier:  Wilmad Labglass ML1451T762
Encompass_Preferred
Separatory Funnel, Squibb, PTFE Stopcock. Capacity: 125mL. Lower Joint Size: [ST] 19/22. Stopcock Size: 2. Stopper Size: 14/20. Supplied complete with glass Pennyhead Stopper of size listed.
Catalog No. 50-804-480