Learn More
Aat Bioquest Fura-10 potassium salt (5x50 ug)
SDP
Supplier:  Aat Bioquest 21110
Encompass_Preferred
In contrast to single-wavelength indicators such as Fluo-4 the absorption or fluorescence excitation maximum of Fura indicators shifts from 380 nm Fura-2 415 nm Fura-8 and Fura-10 for the Ca2-free chelator to about 340 nm Fura-2 355 nm Fura-8 and Fura-10 for the Ca2-bound . This catalog is for 5x50 ug.
Catalog No. 50-224-9969