Learn More
Enzo Life Sciences FURA-2 AM (ultra pure) (1mg). CAS: 108964-32-5
SDP
Supplier:  Enzo Life Sciences ENZ52006
Encompass_Preferred
Fura-2 acetomethoxy (AM) is one of the most widely used calcium indicators for homogenous quantitative dual wavelength ratiometric cell measurements. Fura-2 AM is particularly useful for digital imaging microscopy. It is less susceptible to photobleaching than Indo-1. The probe is excited only by UV light, which results in significantly less interference by visible wavelength excitable fluorescent compounds. One application of the probe is as a useful counter screen tool for GPCR and calcium channel hits identified using calcium indicators excited at 488nm in primary screens. Fura-2 AM can be easily loaded into cells by passive incubation. Wavelength Maxima: Excitation 370nm, Emission 476nm Alternative name: 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2'-amino-5'-methyl-phenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethyl ester. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-2605