Learn More
Cayman Chemical Fura-2potasIum salt 5mg
SDP
Supplier:  Cayman Chemical 195315 MG
Encompass_Preferred
A ratiometric fluorescent calcium indicator; em = 510 nm; ex = 340 and 380 nm at high and low calcium concentrations, respectively; enables determination of ratiometric measurements of calcium influx in live cells
Catalog No. 50-298-2955