Learn More
Aat Bioquest Fura Red AM CAS 149732-62-7 (1 mg)
SDP
Supplier:  Aat Bioquest 21046
Encompass_Preferred
Fura Red is a visible light-excitable fura-2 analog that offers unique possibilities for ratiometric measurement of calcium ion in single cells by microphotometry imaging or flow cytometry when used with single excitation green-fluorescent calcium indicators. This catalog is for 1 mg.
Catalog No. 50-224-9930