Learn More
Apexbio Technology LLC Furaltadone HCl 3759-92-0 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B17645.1
Encompass_Preferred
Furaltadone HCl (CAS 3759-92-0) is a nitrofuran derivative with antibacterial activity Furaltadone HCl exerts its biological effects primarily through interference with bacterial enzymatic pathways resulting in inhibition of microbial growth and replication In in vitro studies Furaltadone HCl demonstrates inhibitory activity against various microbial pathogens with IC50 values varying depending on microbial species and experimental conditions Based on these pharmacological properties Furaltadone HCl holds research potential in the investigation of microbial and protozoal infections particularly in studies of antibacterial and antiprotozoal mechanisms
Catalog No. 50-000-94559