Learn More
Medchemexpress LLC Furaltadone hydrochloride | 3759-92-0 | 99.5% | 1 G
Supplier: Medchemexpress LLC HYB11481G
Furaltadone hydrochloride is an orally active antibacterial agent used for the study of intestinal Salmonella infections. It demonstrates in vitro inhibitory effects on Staphylococcus and shows inhibitory effects on allergic reactions. This compound is suitable for various biological studies.
- Inhibits antigen-induced mast cell degranulation.
- Suppresses the Lyn/Syk signaling pathway.
- Inhibits phosphorylation of early signaling proteins.
- Reduces allergic response in IgE-mediated PCA in mice.