Learn More
Cayman Chemical FuregrelatesodIum salt 10mg
SDP
Supplier:  Cayman Chemical 7054010 MG
Encompass_Preferred
Furegrelate is a potent inhibitor of thromboxane synthase with little effect on other enzymes essential for arachidonate metabolism. The IC50 value is 15 nM for human platelet microsomal thromboxane synthase.{1227,1228}
Catalog No. 50-289-3522