Learn More
Bachem Furin Inhibitor II Trifluoroacetate
SDP
Supplier:  Bachem 4061351.0001
Encompass_Preferred
Furin Inhibitor II Trifluoroacetate, H-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-NH2, 673202-67-0, C36H75N25O6, Mr 954.16, 1mg. Hexa-D-arginine amide. Hexa-D-arginine amide is a potent furin inhibitor with a Ki of 0.106 μM. It also inhibited the related convertases PACE4 and prohormone convertase-1 (PC1) with Ki values of 0.58 μM and 13 μM, respectively. The stable hexapeptide exhibited protection against anthrax toxemia in vitro and in vivo.
Catalog No. NC1859537